Learn More
RAB27B Mouse anti-Human, Unconjugated, Clone: 2B9G9, Proteintech
Código de producto. 16889255 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16889255 20 μL 20 microlitros
16879255 150 μL 150 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16889255 Proveedor Proteintech N.º de proveedor 669441IG20UL
Mouse Monoclonal Antibody
The RAB27B antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RAB27B. This antibody recognizes human antigen. The RAB27B antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Especificaciones
Antígeno RAB27B
Aplicaciones Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
Clasificación Monoclonal
Clon 2B9G9
Concentración 1000 μg/mL
Conjugado Unconjugated
Especie del huésped Mouse
Cantidad 20 μL
Primario o secundario Primary
ID de gen (Entrez) 5874
Especies diana Human
Isotype IgG2a