missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAB21 Polyclonal Antibody

Descripción
A suggested positive control is mouse kidney tissue lysate. PA5-34404 can be used with blocking peptide PEP-1446.
RAB21 is a member of a subfamily of small GTP-binding protein genes of the Ras superfamily that plays an important role in intracellular vesicular targeting. In non-polarized Caco-2 cells, RAB21 showed a general endoplasmic reticulum (ER)-like localization, while in polarized cells, RAB21 localized to apical vesicles. RAB21 has been shown to associate with integrin subunits and to be important for receptor entry into cells via RAB5/RAB21 endosomes. The GTPase-activating protein p120RasGAP regulates cell motility by controlling the return of the endocytosed integrins to the plasma membrane by competing with RAB21 for binding to overlapping sites on the Alpha-tail of endocytosed integrin.
Especificaciones
Especificaciones
| Antígeno | RAB21 |
| Aplicaciones | Immunohistochemistry, Western Blot |
| Clasificación | Polyclonal |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 0.02% sodium azide |
| génica | RAB21 |
| N.º de referencia del gen | P35282, Q6AXT5, Q9UL25 |
| Alias de gen | 9630024B22; GTP-binding protein RAB21; KIAA0118; Rab-12; Rab21; RAB21, member RAS oncogene family; ras-related protein Rab-21 |
| Símbolos de los genes | RAB21 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?