Learn More
Rab18 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Código de producto. 16878283 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16878283 150 μL 150 microlitros
16888283 20 μL 20 microlitros
2 options
Código de producto. 16878283
Marca: Proteintech 113041AP150UL
Rabbit Polyclonal Antibody
The protein encoded by this gene is a member of a family of Ras-related small GTPases that regulate membrane trafficking in organelles and transport vesicles. Knockdown studies is zebrafish suggest that this protein may have a role in eye and brain development. Mutations in this gene are associated with Warburg micro syndrome type 3. Alternatively spliced transcript variants have been found for this gene.
Especificaciones
Antígeno Rab18
Aplicaciones Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Clasificación Polyclonal
Concentración 0.13 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
génica RAB18
N.º de referencia del gen P35293, Q5EB77, Q9NP72
Alias de gen RAB18, RAB18LI1, Ras related protein Rab 18
Símbolos de los genes Rab18
Especie del huésped Rabbit
Inmunógeno Rab18 Fusion Protein Ag1841
Método de purificación Antigen Affinity Chromatography
Cantidad 150 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 19330, 22931, 307039
Especies diana Human, Mouse, Rat
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG