Learn More
R&D Systems™ TACS TdT In Situ Apoptosis Detection Kit, Blue Label
Código de producto. p-4830554
Change view
Click to view available options
Método de detección:
DAB
Etiqueta azul
Tamaño de la unidad:
30 tests
Código de producto. Método de detección unitSize
18432620 DAB 30 tests
18411560 Etiqueta azul 30 tests
2 options
Código de producto. 18432620
Marca: R&D Systems™ 481030K
Includes
Proteinase K Solution; Cytonin™; TdT enzyme, dNTP mix and buffers; Strep-HRP; TACS-Blue Label; Blue Streptavidin-Diluent; Cations; TACS-Nuclease™ and buffer; Nuclear Fast Red
A TUNEL assay designed for fixed cells, embedded tissue, and frozen tissue using light and electron microscopy
Contains a highly purified form of the TdT enzyme for the enzymatic incorporation of biotinylated nucleotides
Biotin labeling is achieved using streptavidin-horseradish peroxidase and the colorimetric substrate TACS Blue Label
TRUSTED_SUSTAINABILITY
Especificaciones
Cantidad 30 Tests
Contenido del kit TUNEL for fixed cells, embedded tissue, frozen tissue using light and electron microscopy; Reagents to detect DNA fragmentn in situ
Tipo de producto TACS TdT In Situ Apoptosis Detection Kit
Contenido y almacenamiento The product requires storage at -20 to -70° C, 2 - 8° C, and room temperature. Consult the product insert for specific storage temperatures. Do not use past expiration date.
Método de detección DAB