Learn More
R&D Systems™ Proteome Profiler Human Protease Inhibitor Array Kit
GREENER_CHOICE
Código de producto. 18017631
Click to view available options
Cantidad:
1 kit
Tamaño de la unidad:
1 set
Código de producto. 18017631
Marca: R&D Systems™ ARY023
Rapid, sensitiv, and economical tool to simultaneously detect the relative changes of 32 protease inhibiting analytes in a single sample
The Proteome Profiler Human Protease Inhibitor Array Kit contains 4 membranes - each spotted in duplicate with 32 different protease inhibitor antibodies.
TRUSTED_SUSTAINABILITY
Especificaciones
Contenido y almacenamiento Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Cantidad 1 kit