Learn More
R&D Systems™ MagCellect Mouse Naive CD4+ T Cell Isolation Kit
Código de producto. 18491980
Click to view available options
Cantidad:
1 kit
Tamaño de la unidad:
1 set
Código de producto. 18491980
Marca: R&D Systems™ MAGM205
Designed to isolate Naive CD4+ T cells via a negative selection principle
CD4 is an approximately 55kDa type I transmembrane glycoprotein that is expressed predominantly on thymocytes and a subset of mature T cells. In humans, CD4 is also expressed to a lesser extent on monocytes and macrophage related cells.
Typical recoveries range between 5% and 35% and the purity of recovered cells ranges between 85 and 95%
Kit protocols based on the use of Ferrofluids or magnetic nanoparticles that have no magnetic memory (superparamagnetic)
These Ferrofluids have a diameter of 150nm and as a result behave like colloidal particles, offering a higher ligand capacity per mass compared to other, larger magnetic particles
TRUSTED_SUSTAINABILITY
Especificaciones
Contenido y almacenamiento Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Cantidad 1 kit