Learn More
R&D Systems™ Human Mesenchymal Stem Cell Functional Identification Kit
Click to view available options
Cantidad:
1 Kit
Tamaño de la unidad:
1 set
Descripción
Includes
Adipogenic Supplement, Osteogenic Supplement, Chondrogenic Supplement, ITS Supplement, Goat anti-mouse FABP-4, Mouse anti-human Osteocalcin, Goat anti-human Aggrecan
Can be used with the Human MSC Verification Flow Kit (Mfr. No. FMC020) to define MSCs according to the ISCT recommendations
Reliably induces MSC trilineage differentiation using defined supplements
Includes premium quality antibodies to confirm differen
Especificaciones
Especificaciones
| Cantidad | 1 Kit |
| Contenido y almacenamiento | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |