Learn More
Quickfit™ Columna de fraccionamiento Dufton de vidrio de borosilicato
Description
- Glass column with accurately fitted Dufton glass spiral
- Good efficiency - liquid spiralling down column meets ascending vapour
- Spiral can be removed for cleaning
Specifications
Specifications
| Longitud (métrico) | 290 mm |
| Tipo | Dufton Fractionating Column |
| Material | Vidrio de borosilicato |
| Certificaciones/conformidad | ISO 3585 |