Learn More
Quercetin, MedChemExpress
Código de producto. p-200096799
Encompass_Preferred
Change view
Click to view available options
Cantidad:
1 g
5 g
Tamaño de la unidad:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16491719 1 g 1 g
16499937 5 g 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16491719 Proveedor Medchem Express N.º de proveedor HY18085
MedChemExpress Quercetin, a natural flavonoid, is a stimulator of recombinant SIRT1 and also a PI3K inhibitor with IC50 of 2.4 μM, 3.0 μM and 5.4 μM for PI3K γ, PI3K δ and PI3K β, respectively.
Quercetin, a natural flavonoid, is a stimulator of recombinant SIRT1 and also a PI3K inhibitor with IC50 of 2.4 μM, 3.0 μM and 5.4 μM for PI3K γ, PI3K δ and PI3K β, respectively.
Especificaciones
Nombre del producto químico o material Quercetin
CAS 117-39-5
Color Yellow
Notas de grado de pureza Research
Fórmula molecular C15H10O7
Cantidad 1 g
Información de solubilidad DMSO : ≥ 100 mg/mL (330.86 mM) ∣Ethanol : 16.67 mg/mL (55.15 mM; Need ultrasonic)
SMILES OC1=C(C2=CC=C(O)C(O)=C2)OC3=CC(O)=CC(O)=C3C1=O
Peso molecular (g/mol) 302.24
Formula Weight (peso de la fórmula) 302.24
Porcentaje de pureza 96.84%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Cancer-Kinase/protease