Learn More
Skytec™ Quartz Safety Gloves
Descripción
- Lightweight with a Breathable PU Palm
- Abrasion and tear resistant with a secure grip
- Thin coating facilitates movement
- Protects from oils and grease
- Elasticated wrist provides a secure
- Lint-free
Especificaciones
Especificaciones
| Grado de abrasión ANSI | ANSI 4 |
| Certificaciones/conformidad (Europa) | CE II, UKCA, ANSI/ISEA 105-2016: Abrasion 4,Puncture 2, EN 388:2016 4131X, EN 388:2016 + A1:2018 4131X |
| Estructura | Seamless Knit |
| Coating Material | Polyurethane |
| Color | White |
| Acabado exterior | No Texture |
| Hand | Ambidextrous |
| Latex Free | Yes |
| Material | Nylon |
| Tipo de producto | General Purpose Gloves |
| Mostrar más |