Learn More
Macherey-Nagel™ Quantofix™ Test Strips
Perform fast, easy analyses on the spot. Macherey-Nagel™ Quantofix™ Test Strips are ready to use semi-quantitative tests that change color depending on the concentration of the analyte in the sample. Read results using the color scale or with the Quantofix™ Relax.
Marca: Macherey-Nagel™ 913918
Descripción
Perform fast, easy analyses on the spot. Macherey-Nagel™ Quantofix™ Test Strips are ready to use semi-quantitative tests that change color depending on the concentration of the analyte in the sample. Read results using the color scale or with the Quantofix™ Relax.
Quantofix™ test strips meet all requirements of a modern rapid test. Analytical professionals as well as occasional testers appreciate the test strips for the fast and easy analysis directly at the point of interest. Often, these tests are used to quickly check whether important parameters are in the desired range. They deliver an immediate result and thus enable a fast response.
- Rapid
- Just Dip & Read
- Results within seconds
- Always ready for use
- Easy
- No calibration
- No maintenance
- No accessories
- Reliable
- Desiccant in the stopper for optimal protection of the strips against humidity
- Color chart confirmed with traceable standards
- Automatic evaluation with Quantofix™ Relax for safe documentation
Especificaciones
| White to Red-Violet | |
| Nitrate/Nitrite | |
| 10 to 500mg/L NO3-, 1-80mg/L NO2- | |
| 0, 10, 25, 50, 100, 250, 500mg/L NO3- 0, 1, 5, 10, 20, 40, 80mg/L NO2- |
| Nitrate/Nitrite | |
| Determination of nitrate | |
| 50 Pack | |
| 60 sec. |