Learn More
Cole-Parmer PVDF Tubing
Código de producto. 30054003 Tienda Cole-Parmer Productos
Change view
Click to view available options
Diámetro interior (imperial):
1/16 in
1/8 in
3/16 in
5/32 pulg
Diámetro exterior (imperial):
1/4 in
1/8 in
3/16 in
Tamaño de la unidad:
15 metros
Código de producto. Diámetro interior (imperial) Diámetro exterior (imperial) unitSize
30054003 1/8 in 3/16 in 15 metros
30053486 5/32 pulg 1/4 in 15 metros
30053793 1/16 in 1/8 in 15 metros
30053029 3/16 in 1/4 in 15 metros
30053399 1/8 in 1/4 in 15 metros
5 options
Código de producto. 30054003
Marca: Cole-Parmer 5011927
Solo quedan null
Añadir a la cesta
Añadir a la cesta
Chemical- and abrasion-resistant tubing at a low cost
Resilient with good fle in., ibility. Does not contain BPA, phthalates, or PVC. Sterilize by autoclave, gamma irradiation, EtO, and chemical sterilants. USP Class VI compliant. Ideal for general-purpose, laboratory, and single-use applications. Free technical application support available to assist with tubing selection
Especificaciones
Certificaciones/conformidad ADCF;Non-Phthalate;RoHS;USP
Diámetro interior (imperial) 1/8 in
Diámetro exterior (imperial) 3/16 in
Longitud global (imperial) 50 ft.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.