Learn More
- Home
- Products
- Tubos
- Tubos de silicona y caucho
- Cole-Parmer PVC Lab and Vacuum Tubing
Cole-Parmer PVC Lab and Vacuum Tubing
Código de producto. 30054239 Tienda Cole-Parmer Productos
Change view
Click to view available options
Diámetro interior (imperial):
1/16 in
1/2 in
1/4 in
1/5 in
1/8 in
15/64 pulg
1 in
3/16 in
3/32 in
3/4 in
3/8 in
5/16 in
5/32 pulg
5/64 in
Diámetro exterior (imperial):
1-1/4 in
1-1/8 in
1/2 in
1/4 in
1/5 in
1/8 in
11/32 in
15/32 in
15/64 in
1 in
3/16 in
3/4 in
3/8 in
5/16 in
5/32 pulg
5/8 in
7/16 in
7/8 in
9/16 in
Tamaño de la unidad:
15 metros
| Código de producto. | Diámetro interior (imperial) | Diámetro exterior (imperial) | unitSize |
|---|---|---|---|
| 30054239 | 5/64 in | 5/32 pulg | 15 metros |
| 30054031 | 3/8 in | 1/2 in | 15 metros |
| 30053338 | 1/16 in | 1/8 in | 15 metros |
| 30053611 | 1/4 in | 5/8 in | 15 metros |
| 30053704 | 1/2 in | 5/8 in | 15 metros |
| 30053850 | 1/2 in | 3/4 in | 15 metros |
| 30053084 | 3/32 in | 5/32 pulg | 15 metros |
| 30053748 | 3/8 in | 7/8 in | 15 metros |
| 30054309 | 1/2 in | 1-1/8 in | 15 metros |
| 30053306 | 3/8 in | 9/16 in | 15 metros |
| 30053415 | 1/8 in | 1/4 in | 15 metros |
| 30053287 | 3/4 in | 1 in | 15 metros |
| 30053926 | 1 in | 1-1/4 in | 15 metros |
| 30053701 | 1/4 in | 3/8 in | 15 metros |
| 30052936 | 5/16 in | 1/2 in | 15 metros |
| 30053886 | 5/16 in | 7/16 in | 15 metros |
| 30053433 | 3/16 in | 9/16 in | 15 metros |
| 30053381 | 1/8 in | 3/16 in | 15 metros |
| 30053136 | 3/8 in | 5/8 in | 15 metros |
| 30053449 | 5/32 pulg | 15/64 in | 15 metros |
| 30053173 | 1/8 in | 1/5 in | 15 metros |
| 30054048 | 1/5 in | 5/16 in | 15 metros |
| 30054189 | 15/64 pulg | 11/32 in | 15 metros |
| 30052944 | 1/4 in | 7/16 in | 15 metros |
| 30053670 | 5/16 in | 15/32 in | 15 metros |
Este artículo no se puede devolver.
Vea la política de devoluciones
Código de producto. 30054239
Marca: Cole-Parmer 5012109
Inicie sesión o regístrese para comprobar su precio y disponibilidad.
Añadir a la cesta
Añadir a la cesta
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Versatile applications including peristaltic pumps, analytical instruments, incubators, gas, and drain lines. Glass-smooth inner bore design for easy clean-ups and changeovers, enhancing workflow efficiency. Ideal for general-purpose, laboratory, and single-use applications. Free technical application support available to assist with tubing selection
Especificaciones
Especificaciones
| Diámetro interior (imperial) | 5/64 in |
| Material | PVC |
| Diámetro exterior (imperial) | 5/32 pulg |
| Longitud global (imperial) | 50 ft. |
| Color | Translúcido |
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.