Learn More
PURA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Código de producto. 16887684 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16887684 150 μL 150 microlitros
16897684 20 μL 20 microlitros
2 options
Código de producto. 16887684
Marca: Proteintech 177331AP150UL
Rabbit Polyclonal Antibody
This gene product is a sequence-specific, single-stranded DNA-binding protein. It binds preferentially to the single strand of the purine-rich element termed PUR, which is present at origins of replication and in gene flanking regions in a variety of eukaryotes from yeasts through humans. Thus, it is implicated in the control of both DNA replication and transcription. Deletion of this gene has been associated with myelodysplastic syndrome and acute myelogenous leukemia.
Especificaciones
Antígeno PURA
Aplicaciones Western Blot
Clasificación Polyclonal
Concentración 0.21 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica PURA
N.º de referencia del gen P42669, P86252, Q00577
Alias de gen PUR ALPHA, PUR1, PURA, PURALPHA
Símbolos de los genes PURA
Especie del huésped Rabbit
Inmunógeno PURA Fusion Protein Ag12173
Método de purificación Antigen Affinity Chromatography
Cantidad 150 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 19290, 307498, 5813
Especies diana Human, Mouse, Rat
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG