missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTPN14 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 18053-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP contains an N-terminal noncatalytic domain similar to that of band 4.1 superfamily cytoskeleton-associated proteins, which suggested the membrane or cytoskeleton localization of this protein. The specific function of this PTP has not yet been determined.Especificaciones
| PTPN14 | |
| Polyclonal | |
| Unconjugated | |
| PTPN14 | |
| PEZ, PTP36, PTPN14 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 19250, 305064, 5784 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q15678, Q62130 | |
| PTPN14 | |
| PTPN14 Fusion Protein Ag12571 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido