missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PTPN13 Polyclonal Antibody
GREENER_CHOICE

Código de producto. 17814380
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
17814380 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 17814380 Proveedor Invitrogen™ N.º de proveedor PA5139860

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: STPVHLTNEM KNYMKKSSYM QDSAIDSSSK DHHWSRGTLR HISENSFGPS GGLREGSLSS QDSRTESASL SQSQVNGFFA SHLGDQTWQE SQHGSPSPSV ISKATE Highest antigen sequence indentity to the following orthologs: Mouse - 75%, Rat - 73%.

The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP is a large intracellular protein. It has a catalytic PTP domain at its C-terminus and two major structural domains: a region with five PDZ domains and a FERM domain that binds to plasma membrane and cytoskeletal elements. This PTP was found to interact with, and dephosphorylate, Fas receptor and IkappaBalpha through the PDZ domains. This suggests it has a role in Fas mediated programmed cell death. This PTP was also shown to interact with GTPase-activating protein, and thus may function as a regulator of Rho signaling pathways. Four alternatively spliced transcript variants, which encode distinct proteins, have been reported.
TRUSTED_SUSTAINABILITY

Specifica

Antígeno PTPN13
Aplicaciones Immunocytochemistry
Clasificación Polyclonal
Concentración 0.2 mg/mL
Conjugado Unconjugated
Formulación PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
génica PTPN13
N.º de referencia del gen Q12923
Alias de gen AI324989; DKFZp686J1497; FAP-1; Fas-associated protein-tyrosine phosphatase 1; hPTP1E; hPTPE1; PNP1; protein Tyr phosphatase; protein tyrosine phosphatase DPZPTP; Protein tyrosine phosphatase PTP-BL; protein tyrosine phosphatase, non-receptor type 13; protein tyrosine phosphatase, non-receptor type 13 (APO-1/CD95 (Fas)-associated phosphatase); protein-tyrosine phosphatase 1E; Protein-tyrosine phosphatase PTPL1; protein-tyrosine phosphatase RIP; ptp; Ptp14; PTP1E; PTP36; PTP-BAS; PTP-BL; PTP-E1; PTPL1; PTPLE; PTPN13; Ptpri; RIP; tyrosine phosphatase; tyrosine-protein phosphatase non-receptor type 13
Símbolos de los genes PTPN13
Especie del huésped Rabbit
Inmunógeno Recombinant protein corresponding to Human PTPN13. Recombinant protein control fragment (Product #RP-108381).
Método de purificación Antigen affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 5783
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.