missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PTBP1 Polyclonal Antibody
GREENER_CHOICE

Código de producto. 16332905
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
Código de producto. Cantidad unitSize
16332905 100 μL 100 microlitros
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 16332905

Marca: Invitrogen™ PA585996

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Rabbit Polyclonal Antibody

PTBP1 Polyclonal Antibody for ICC/IF, IHC (P), Western Blot

This gene belongs to the subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs). The hnRNPs are RNA-binding proteins and they complex with heterogeneous nuclear RNA (hnRNA). These proteins are associated with pre-mRNAs in the nucleus and appear to influence pre-mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. The protein encoded by this gene has four repeats of quasi-RNA recognition motif (RRM) domains that bind RNAs. This protein binds to the intronic polypyrimidine tracts that requires pre-mRNA splicing and acts via the protein degradation ubiquitin-proteasome pathway. It may also promote the binding of U2 snRNP to pre-mRNAs. This protein is localized in the nucleoplasm and it is also detected in the perinucleolar structure. Alternatively spliced transcript variants encoding different isoforms have been described.
TRUSTED_SUSTAINABILITY

Spécification

Antígeno PTBP1
Aplicaciones Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Clasificación Polyclonal
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 0.2% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
génica PTBP1
N.º de referencia del gen P17225, P26599, Q00438
Alias de gen 57 kDa RNA-binding protein PPTB-1; AA407203; AL033359; Heterogeneous nuclear ribonucleoprotein I; heterogeneous nuclear ribonucleoprotein polypeptide I; hnRNP I; HNRNPI; HNRNP-I; HNRPI; MGC10830; MGC8461; polypyrimidine tract binding protein (heterogeneous nuclear ribonucleoprotein I); polypyrimidine tract binding protein 1; polypyrimidine tract-binding protein; polypyrimidine tract-binding protein 1; polypyrimidine-tract binding protein; pPTB; PTB; PTB-1; PTB2; PTB3; PTB4; PTBP1; PTB-T; Pybp; Pybp1; Pybp2; pyrimidine binding protein 1; pyrimidine binding protein 2; RNA-binding protein
Símbolos de los genes PTBP1
Especie del huésped Rabbit
Inmunógeno Synthetic peptide corresponding to Human PTBP1 aa 1-50.
Método de purificación Antigen affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 19205, 29497, 5725
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.