Learn More
Invitrogen™ PSPN Monoclonal Antibody (275508)

Mouse Monoclonal Antibody
Marca: Invitrogen™ MA523978
Descripción
In Western blots, no cross-reactivity with recombinant human (rh) AgRP, rhArtemin, rh beta IG-H3, rhCripto-1, recombinant drosophila Decapentaplegic, rhGDNF, rhLAP, rhLatent TGF-beta 1, rhLefty-A, rhMIS, rhNeurturin, recombinant mouse (rh) Nodal, rmPersephin, rhTGF-alpha, rhTGF-beta 1, rhTGF-beta 1.2, rhTGF-beta 2, rhTGF-beta 3, or recombinant amphibian TGF-beta 5 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
Persephin is a disulfide-linked, homodimeric, neurotrophic factor structurally related to GDNF, artemin, and neurturin. These proteins belong to the cysteine-knot family of growth factors that assume stable dimeric structures. Persephin signals through a multicomponent receptor system, composed of RET and one of four GFR alpha (alpha1-alpha4) receptors. The GFRalpha4 was first identified in chicken, and was later shown to be the preferential binding subunit for persephin. Persephin promotes the survival of ventral midbrain dopaminergic neurons and motor neurons after sciatic nerve oxotomy, and, like GDNF, promotes ureteric bud branching. However, in contrast to GDNF and neurturin, persephin does not support the survival of peripheral neurons.
Especificaciones
| PSPN | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| O60542 | |
| PSPN | |
| E. coli-derived recombinant human Persephin Ala61-Gly156. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry, Western Blot | |
| 275508 | |
| Unconjugated | |
| PSPN | |
| Persephin; PSP; Pspn | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 5623 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.