missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PSMD6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA5118212
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
PSMD6 Polyclonal Antibody for Western Blot, IP
This gene encodes a member of the protease subunit S10 family. The encoded protein is a subunit of the 26S proteasome which colocalizes with DNA damage foci and is involved in the ATP-dependent degradation of ubiquinated proteins. Alternative splicing results in multiple transcript variants.
Especificaciones
| PSMD6 | |
| Polyclonal | |
| PBS with 0.03% ProClin 300; pH 7 | |
| Q15008 | |
| PSMD6 | |
| E. coli-derived Human PSMD6 fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| PSMD6 | |
| 2400006A19Rik; 26S proteasome non-ATPase regulatory subunit 6; 26S proteasome non-ATPase regulatory subunit 6-like protein; 26S proteasome non-ATPase regulatory subunit-like protein; 26S proteasome regulatory subunit RPN7; 26S proteasome regulatory subunit S10; breast cancer-associated protein SGA-113M; KIAA0107; p42A; P44s10; PFAAP4; Phosphonoformate immuno-associated protein 4; proteasome (prosome, macropain) 26S subunit, non-ATPase, 6; proteasome 26S subunit, non-ATPase 6; proteasome regulatory particle subunit p44S10; proteasome, 26S, non-ATPase regulatory subunit 6; PSMD6; Rpn7; S10; SGA-113M; vov; zgc:56471 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 9861 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido