Learn More
Invitrogen™ PSGR Polyclonal Antibody
Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA5104387
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Antibody detects endogenous levels of total OR51E2.
PSGR (prostate-specific G-protein coupled receptor), also known as OR51E2 (olfactory receptor 51E2), is a 320 amino acid multi-pass membrane protein that belongs to the olfactory receptor subfamily of G-protein coupled receptors. Expressed exclusively in prostate tissue and upregulated in prostate cancer, PSGR functions as an odorant receptor that binds odorant molecules and triggers the perception of smell.
Especificaciones
| PSGR | |
| Polyclonal | |
| Unconjugated | |
| OR51E2 | |
| 4633402A21Rik; GA_x6K02T2PBJ9-5459657-5458695; G-protein coupled receptor RA1c; hCG_2044604; HPRAJ; Mol2.3; MOR18-2; olfactory receptor 51E2; Olfactory receptor 59; olfactory receptor 78; olfactory receptor family 51 subfamily E member 2; olfactory receptor MOR18-2; olfactory receptor OR11-16; olfactory receptor, family 51, subfamily E, member 2; olfactory receptor, family 51, subfamily E, member 3 pseudogene; olfactory receptor, family 52, subfamily A, member 2; Olfr78; Olr59; OR51E2; OR51E3P; OR52A2; prostate specific G-protein coupled receptor; prostate-specific G-protein coupled receptor; PSGR; RA1c | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 170816, 81285 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O88628, Q9H255 | |
| Olr59, OR51E2 | |
| A synthesized peptide derived from human OR51E2(Accession Q9H255), corresponding to amino acid residues L204-L254. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |