missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRX Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA579887
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human Jurkat whole cell, human K562 whole cell, humna U2OS whole cell, human U251 whole cell, rat brain tissue, mouse brain tissue. Flow: K562 cell.
Periaxin is a protein that in humans is encoded by the PRX gene. This gene encodes a protein involved in peripheral nerve myelin upkeep. The encoded protein contains 2 PDZ domains which were named after PSD95 (post synaptic density protein), DlgA (Drosophila disc large tumor suppressor), and ZO1 (a mammalian tight junction protein). Two alternatively spliced transcript variants have been described for this gene which encode different protein isoforms and which are targeted differently in the Schwann cell. Mutations in this gene cause Charcot-Marie-Tooth neuoropathy, type 4F and Dejerine-Sottas neuropathy.
Especificaciones
| PRX | |
| Polyclonal | |
| Unconjugated | |
| PRX | |
| CMT4F; KIAA1620; L-Periaxin; periaxin; PRX | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 19153, 57716, 78960 | |
| -20°C | |
| Lyophilized |
| ELISA, Flow Cytometry, Western Blot, Western Blot | |
| 500 μg/mL | |
| PBS with 4mg trehalose and no preservative | |
| O55103, Q63425, Q9BXM0 | |
| PRX | |
| E. coli-derived human PRX recombinant protein (Position: M1-K91). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido