missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Protein S Monoclonal Antibody (5092)

Descripción
MA1-43018 detects Protein S in human samples. MA1-43018 has successfully been used in ELISA, WB (Non-reducing), RIA applications. Ca2+ is required. MA1-43018 was derived from human protein S.
Protein S is a vitamin K-dependent, potent neutral anticoagulant plasma glycoprotein. Protein S has a molecular weight of approximately 70 kDa and consists of a single polypeptide chain. The concentration of protein S in human plasma is 25 μg/mL, with a half-life of approximately two days. About 40% of Protein S in plasma is in a free form, whereas 60% is complexed with C4b-binding Protein (C4BP). Only the free Protein S functions as a cofactor to antigen presenting cells (APC) in the APC-dependent degradation of Factor Va and Factor VIIIa.
Especificaciones
Especificaciones
| Antígeno | Protein S |
| Aplicaciones | ELISA, Radioimmune Assays (RIA), Western Blot |
| Clasificación | Monoclonal |
| Clon | 5092 |
| Concentración | 5.4 mg/mL |
| Conjugado | Unconjugated |
| Formulación | 50% water with 50% glycerol and no preservative |
| génica | PROS1 |
| N.º de referencia del gen | P07225 |
| Alias de gen | AW214361; Preproprotein S; PROS; PROS 1; PROS1; protein S; protein S (alpha); Protein S alpha; protein Sa; proteinS; PS 21; PS 22; PS 23; PS 24; PS 25; PS 26; PS21; PS22; PS23; PS24; PS25; PS26; PSA; rabbit protein S; THPH5; THPH6; unnamed protein product; vitamin K-dependent plasma protein S; vitamin K-dependent protein S; vitamin K-dependent protein S precursor |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?