missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Protein APC Monoclonal Antibody (CC-1), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA125884
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
MA1-25884 detects APC from human, mouse, and rat samples. MA1-25884 has been successfully used in immunohistochemistry (paraffin), immunofluorescence, and immunocytochemsitry procedures. The MA1-25884 immunogen is recombit N-terminal fragment of APC.
This gene encodes a tumor suppressor protein that includes among its many intracellular functions one of nuclear export. Defects in this gene cause familial adenomatous polyposis (FAP), an autosomal domit pre-maligt disease that usually progresses to maligcy. Disease-associated mutations tend to be clustered in a small region designated the mutation cluster region (MCR) and result in a truncated protein product.
Especificaciones
| Protein APC | |
| Monoclonal | |
| 0.1 mg/mL | |
| APC | |
| Adenomatous Polyposis Coli, APC, DP2, DP2.5, DP3, FAP, FPC, GS | |
| Mouse | |
| Antigen affinity chromatography | |
| RUO | |
| 11789, 24205, 324 | |
| 4° C, do not freeze | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
| CC-1 | |
| Unconjugated | |
| P25054, Q61315, P70478 | |
| APC | |
| Recombit N-terminal fragment of APC. | |
| 50 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2b |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido