missing translation for 'onlineSavingsMsg'
Learn More

Progesterone Receptor Monoclonal Antibody (hPRa 7), Invitrogen™

Código de producto. 11539830
Change view
Click to view available options
Cantidad:
500 μL
Tamaño de la unidad:
500 microlitros
Código de producto. Cantidad unitSize
11539830 500 μL 500 microlitros
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 11539830

Marca: Invitrogen MA512658

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

MA5-12658 targets Progesterone Receptor in IF, IP, and WB applications and shows reactivity with Human, mouse, Non-human primate, and Rat samples. The MA5-12658 immunogen is pgR from a human endometrial carcinoma (EnCa 101) grown in athymic mice.

The progesterone receptor (PR) is a member of the steroid family of nuclear receptors. The PR mediates the physiological effects of progesterone, which plays a central role in reproductive events associated with the establishment and maintenance of pregnancy. PR is found as a 94 kDa protein (Form A) or a 120 kDa protein (Form B) due to the use of alternative translation initiation sites. PR-B is the transcriptionally active form and is responsible for activating genes for the maintenance of the endometrium, maintenance of pregnancy, and inhibition of ovulation. PR-A is identical to PR-B except for a 165 amino acid deletion at the N-terminus. This deletion exposes a 140 amino acid inhibitory domain (ID) that acts as a repressor of steroid hormone transcriptional activity. In its inactive state, PgR forms a multiprotein complex which includes heat shock proteins and immunophins. Upon binding of progesterone hormone to its receptor, there is a conformational change that allows dimerization and binding of the receptor to progesterone response elements (PRE) sequences, resulting in activated transcription. A Null mutation in the PGR gene leads to pleiotrophic reproductive abnormalities.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno Progesterone Receptor
Aplicaciones Immunocytochemistry, Immunoprecipitation, Western Blot
Clasificación Monoclonal
Clon hPRa 7
Concentración 0.2 mg/mL
Conjugado Unconjugated
Formulación PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
génica PGR
N.º de referencia del gen P06401, Q00175, Q63449
Alias de gen 9930019P03Rik; BB114106; ENSMUSG00000074510; LOW QUALITY PROTEIN: progesterone receptor; NR3C3; Nuclear receptor subfamily 3 group C member 3; p4 receptor; PGR; pot. alt. progesterone receptor; putative; PR; PR-A; PR-B; Progesterone receptor; progesterone receptor A; progesterone-receptor; putative form B; steroid hormone receptor; testicular progesterone receptor; unnamed protein product
Símbolos de los genes PGR
Especie del huésped Mouse
Inmunógeno PgR from a human endometrial carcinoma (EnCa 101) grown in athymic mice
Método de purificación Protein G
Cantidad 500 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 18667, 25154, 5241
Especies diana Human, Mouse, Non-human Primate, Rat
Contenido y almacenamiento 4° C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG1 κ
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.