Learn More
Thermo Scientific Chemicals Proctolin
Click to view available options
Cantidad:
1 mg
Tamaño de la unidad:
1 miligramo
Descripción
Proctolin modulates interneuronal and neuromuscular synaptic transmission in a wide variety of arthropods.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Especificaciones
| Método de purificación | HPLC |
| Contenido y almacenamiento | -30°C to -10°C |
| Formato | Polvo |
| Para utilizar con (aplicación) | An insect neuropeptide |
| Peso molecular | 648.76 |
| Tipo de producto | Proctolin |
| Cantidad | 1 mg |
| Estado normativo | RUO - research use only |
| Secuencia | Arg-Tyr-Leu-Pro-Thr |