Learn More
Pro-Lab Diagnostics™ Prolex™ Staph Xtra Latex Kits
Click to view available options
Cantidad:
100 pruebas
Tamaño de la unidad:
paquete de 100
Descripción
Includes
- Staph Xtra Latex Reagent: 2 x 2.5mL (PL.1080), 2 x 7.5mL (PL.1081)
- Negative Control Latex Reagent: 5.0mL (PL.1080), 2 x 7.5mL (PL.1081)
- Agglutination Cards: 20 (PL.1080), 48 (PL.1081)
- Mixing Sticks: 200 (PL.1080), 300 (PL.1081)
- Rapid clear results within 20 seconds
- Uses blue latex particles (CMMP) against a white background
Especificaciones
Especificaciones
| Analitos detectables | S. aureus |
| Tipo | Staph Xtra Latex Kit |
| Incluye | 2 x 2.5mL Staph Xtra Latex reagent, 2.5mL negative control Latex reagent, 20 Agglutination cards, 200 mixing sticks |
| Cantidad | 100 pruebas |