missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRMT5 Mouse anti-Human, Clone: OTI3E6, lyophilized, TrueMAB™
Descripción
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Arginine methylation is an irreversible post translational modification which has only recently been linked to protein activity. At least three types of PRMT enzymes have been identified in mammalian cells. These enzymes have been shown to have essential regulatory functions by methylation of key proteins in several fundamental areas. These protein include nuclear proteins, IL enhancer binding factor, nuclear factors, cell cycle proteins, signal transduction proteins, apoptosis proteins, and viral proteins. The mammalian PRMT family currently consists of 7 members that share two large domains of homology. Outside of these domains, epitopes were identified and antibodies against all 7 PRMT members have been developed.
Especificaciones
Especificaciones
| Antígeno | PRMT5 |
| Aplicaciones | Western Blot |
| Clasificación | Monoclonal |
| Clon | OTI3E6 |
| Conjugado | Unconjugated |
| Formulación | PBS with 8% trehalose and no preservative; pH 7.3 |
| génica | PRMT5 |
| N.º de referencia del gen | O14744 |
| Alias de gen | HRMT1L5, IBP72, JBP1, SKB1, SKB1Hs |
| Símbolos de los genes | PRMT5 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?