missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Presenilin 1 Recombinant Rabbit Monoclonal Antibody (24GB4515)
GREENER_CHOICE

Código de producto. p-200110959
Change view
Click to view available options
Cantidad:
20 μL
100 μL
Tamaño de la unidad:
100 microlitros
20 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
30276228 100 μL 100 microlitros
30276008 20 μL 20 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 30276228 Proveedor Invitrogen™ N.º de proveedor MA557620

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Recombinant Monoclonal Antibody

Presenilin 1 Recombinant Monoclonal Antibody for Western Blot, Flow

Presenilin1 was initially identified a marker of susceptibility to early-onset Alzheimer's disease. In addition to PEN2, nicastrin and APH-1, Presenilin1 forms the -gamma-secretase protein complex, a membrane-bound aspartyl protease that can cleave certain proteins at peptide bonds buried within the hydrophobic environment of the lipid bilayer. This cleavage is responsible for a key step in signaling from several cell-surface receptors and is thought to be required for the generation of the neurotoxic amyloid peptides that are central to the pathogenesis of Alzheimer's disease. Like the tumor necrosis factor-alpha-converting enzyme (TACE) and the beta-site cleavage enzyme (BACE) protease families, -gamma-secretase will cleave the amyloid precursor protein (APP), but within the intramembrane region of APP, resulting in either the non-toxic p3 (from the alpha and -gamma cleavage site) or the toxic Abeta amyloid peptide (from the beta and -gamma cleavage site). It is thought that accumulation of the Abeta peptide is the precursor to Alzheimer's disease. Multiple isoforms of presenilin1 are known to exist.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno Presenilin 1
Aplicaciones Flow Cytometry, Western Blot, Immunocytochemistry
Clasificación Recombinant Monoclonal
Clon 24GB4515
Concentración 0.2 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
génica PSEN1
N.º de referencia del gen P49768, P49769, P97887
Alias de gen AD3; Ad3h; Alzheimer Disease 3; FAD; Minilin; presenilin 1; presenilin 1 (Alzheimer disease 3); presenilin1; presenilin-1; Presenilin-1 CTF subunit; Presenilin-1 CTF12; Presenilin-1 NTF subunit; Protein S182; PS1; PS-1; PS1-CTF12; PSEN1; Psnl1; S182; S182 protein; Senilin 1
Símbolos de los genes PSEN1
Especie del huésped Rabbit
Inmunógeno A synthesized peptide derived from human Presenilin 1 (aa 346-361).
Método de purificación Affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 19164, 29192, 5663
Especies diana Human, Mouse, Rat
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.