Learn More
DiscoverX PrecisION™ hNav1.3 Recombinant Stable Cell Line
Click to view available options
Cantidad:
2 viales
Tamaño de la unidad:
2 ampollas
Descripción
The PrecisION™ hNav1.3 Recombinant Stable Cell Line is a stable, engineered cell line for use in studying Nav1.3 ion channels, including drug candidates which affect channel function. This cell line enables assessment of ion channel function via detection of electrophysiological signals. The included cell line overexpresses Nav1.3 ion channels. Upon ligand/agonist induced channel activation, response can be detected via automated or manual electrophysiological techniques. This stable modified cell line is provided as two vials of cryopreserved cells.
Especificaciones
Especificaciones
| Entrada del ensayo | Ion Channel Activity |
| Línea de células | CHO-K1 |
| Para utilizar con (aplicación) | Cell Lines |
| Cantidad | 2 viales |
| Entrada diana | Nav1.3 |
| Número de acceso | AJ251507 |