Learn More
Ludwig Schneider Precision ASTM hydrometers, S506HL-14, cal. at 15°C, 900-950 in 0.5 kg/m3L: 380 mm Thermo: -20+65°C
Marca: Ludwig Schneider 3222117S
Precision ASTM hydrometers, S506HL-14, cal. at 15°C, 900-950 in 0.5 kg/m3L: 380 mm Thermo: -20+65°C