missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PREB Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB120-3422
This item is not returnable.
View return policy
Description
PREB Polyclonal specifically detects PREB in Mouse samples. It is validated for Western Blot.
Specifications
| PREB | |
| Polyclonal | |
| Western Blot 1:500 | |
| MGC3467, prolactin regulatory binding-element protein, prolactin regulatory element binding, prolactin regulatory element-binding protein, SEC12Mammalian guanine nucleotide exchange factor mSec12 | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Detects recombinant mouse PREB (Prolactin Regulatory Element Binding protein). | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| PREB | |
| Recombinant mouse PREB. | |
| 100 μL | |
| Transcription Factors and Regulators | |
| 10113 | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction