missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PPP4R4 Polyclonal Antibody

Descripción
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
The protein encoded by this gene is a HEAT-like repeat-containing protein. The HEAT repeat is a tandemly repeated, 37-47 amino acid long module occurring in a number of cytoplasmic proteins. Arrays of HEAT repeats form a rod-like helical structure and appear to function as protein-protein interaction surfaces. The repeat-containing region of this protein has some similarity to the constant regulatory domain of the protein phosphatase 2A PR65/A subunit. The function of this particular gene product has not been determined. Alternative splicing has been observed for this gene and two transcript variants encoding distinct isoforms have been identified.
Especificaciones
Especificaciones
| Antígeno | PPP4R4 |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.5 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 2% sucrose and 0.09% sodium azide |
| génica | PPP4R4 |
| N.º de referencia del gen | Q8C0Y0 |
| Alias de gen | 8430415E04Rik; CFAP14; cilia and flagella associated protein 14; HEAT-like repeat-containing protein; KIAA1622; mKIAA1622; PP4R4; PPP4R4; protein phosphatase 4 regulatory subunit 4; protein phosphatase 4, regulatory subunit 4; RGD1560932; Serine/threonine-protein phosphatase 4 regulatory subunit 4 |
| Símbolos de los genes | PPP4R4 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?