missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PPP2R2A Recombinant Rabbit Monoclonal Antibody (1H3L5)

Código de producto. 15805753
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
Código de producto. Cantidad unitSize
15805753 100 μg 100 microgramos
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 15805753

Marca: Invitrogen™ 701842

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Recombinant Monoclonal Antibody

This antibody is predicted to react with Monkey, Mouse, Pig Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

The product of this gene belongs to the phosphatase 2 regulatory subunit B family. Protein phosphatase 2 is one of the four major Ser/Thr phosphatases, and it is implicated in the negative control of cell growth and division. It consists of a common heteromeric core enzyme, which is composed of a catalytic subunit and a constant regulatory subunit, that associates with a variety of regulatory subunits. The B regulatory subunit might modulate substrate selectivity and catalytic activity. This gene encodes an alpha isoform of the regulatory subunit B55 subfamily. Alternatively spliced transcript variants have been described.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno PPP2R2A
Aplicaciones Western Blot, Immunocytochemistry
Clasificación Recombinant Monoclonal
Clon 1H3L5
Concentración 0.5 mg/mL
Conjugado Unconjugated
Formulación PBS with 0.09% sodium azide; pH 7.4
génica PPP2R2A
N.º de referencia del gen P63151, Q6P1F6
Alias de gen 2410004D02Rik; B55A; B55ALPHA; hypothetical protein; mKIAA1541; PP2A subunit B isoform alpha; PP2A subunit B isoform B55-alpha; PP2A subunit B isoform BRA; PP2A subunit B isoform PR55-alpha; PP2A subunit B isoform R2-alpha; PP2A, subunit B, B55-alpha isoform; PP2A, subunit B, B-alpha isoform; PP2A, subunit B, BRA isoform; PP2A, subunit B, PR55-alpha isoform; PP2A, subunit B, R2-alpha isoform; PPP2R2A; PR52A; PR55A; protein phosphatase 2 (formerly 2A), regulatory subunit B (PR 52), alpha isoform; protein phosphatase 2 (formerly 2A), regulatory subunit B (PR52), alpha isoform; protein phosphatase 2 (formerly 2A), regulatory subunit B, alpha isoform; protein phosphatase 2 regulatory subunit B, alpha; protein phosphatase 2 regulatory subunit Balpha; protein phosphatase 2, regulatory subunit B, alpha; protein phosphatase 2A 55 kDa regulatory subunit, alpha isoform; serine/threonine protein phosphatase 2A, 55 KDA regulatory subunit B, alpha isoform; serine/threonine-protein phosphatase 2A 55 kDa regulatory subunit B alpha isoform; serine-threonine protein phosphatase 2A regulatory subunit; testicular tissue protein Li 156
Símbolos de los genes PPP2R2A
Especie del huésped Rabbit
Inmunógeno Protein corresponding to human PPP2R2A (aa16-aa227).
Método de purificación Protein A
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 5520, 71978
Especies diana Human, Mouse
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.