missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PPP2R2A Polyclonal Antibody
GREENER_CHOICE

Código de producto. 11572410
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
11572410 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 11572410 Proveedor Invitrogen™ N.º de proveedor PA517515

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

It is not recommended to aliquot this antibody. This antibody is not cross-reactive with the B-prime (PR61), B-prime-prime, or B-prime-prime-prime families of PP2A B subunits.

The product of this gene belongs to the phosphatase 2 regulatory subunit B family. Protein phosphatase 2 is one of the four major Ser/Thr phosphatases, and it is implicated in the negative control of cell growth and division. It consists of a common heteromeric core enzyme, which is composed of a catalytic subunit and a constant regulatory subunit, that associates with a variety of regulatory subunits. The B regulatory subunit might modulate substrate selectivity and catalytic activity. This gene encodes an alpha isoform of the regulatory subunit B55 subfamily. Alternatively spliced transcript variants have been described.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno PPP2R2A
Aplicaciones Immunoprecipitation, Western Blot
Clasificación Polyclonal
Concentración 17 μg/mL
Conjugado Unconjugated
Formulación 0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5
génica PPP2R2A
N.º de referencia del gen P36876, P63151, Q6P1F6
Alias de gen 2410004D02Rik; B55A; B55ALPHA; hypothetical protein; mKIAA1541; PP2A subunit B isoform alpha; PP2A subunit B isoform B55-alpha; PP2A subunit B isoform BRA; PP2A subunit B isoform PR55-alpha; PP2A subunit B isoform R2-alpha; PP2A, subunit B, B55-alpha isoform; PP2A, subunit B, B-alpha isoform; PP2A, subunit B, BRA isoform; PP2A, subunit B, PR55-alpha isoform; PP2A, subunit B, R2-alpha isoform; PPP2R2A; PR52A; PR55A; protein phosphatase 2 (formerly 2A), regulatory subunit B (PR 52), alpha isoform; protein phosphatase 2 (formerly 2A), regulatory subunit B (PR52), alpha isoform; protein phosphatase 2 (formerly 2A), regulatory subunit B, alpha isoform; protein phosphatase 2 regulatory subunit B, alpha; protein phosphatase 2 regulatory subunit Balpha; protein phosphatase 2, regulatory subunit B, alpha; protein phosphatase 2A 55 kDa regulatory subunit, alpha isoform; serine/threonine protein phosphatase 2A, 55 KDA regulatory subunit B, alpha isoform; serine/threonine-protein phosphatase 2A 55 kDa regulatory subunit B alpha isoform; serine-threonine protein phosphatase 2A regulatory subunit; testicular tissue protein Li 156
Símbolos de los genes PPP2R2A
Especie del huésped Rabbit
Inmunógeno Synthetic peptide corresponding to amino acids near the amino terminus of human PP2A B subunit.
Método de purificación Antigen affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 117104, 5520, 71978
Especies diana Human, Mouse, Monkey, Rat
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.