missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PPP2R1A Polyclonal Antibody
GREENER_CHOICE

Código de producto. 13401777
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
Código de producto. Cantidad unitSize
13401777 100 μL 100 microlitros
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 13401777

Marca: Invitrogen™ PA527643

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

Recommended positive controls: HeLa, NIH-3T3, JC, BCL-1. Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (88%), Xenopus laevis (95%), Pig (100%), Rhesus Monkey (100%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The PR65 subunit of protein phosphatase 2A serves as a scaffolding molecule to coordinate the assembly of the catalytic subunit and a variable regulatory B subunit. Upon interaction with GNA12 promotes dephosphorylation of microtubule associated protein TAU/MAPT. Required for proper chromosome segregation and for centromeric localization of SGO1 in mitosis.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno PPP2R1A
Aplicaciones Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Clasificación Polyclonal
Concentración 0.12 mg/mL
Conjugado Unconjugated
Formulación PBS with 20% glycerol and 0.025% ProClin 300; pH 7
génica PPP2R1A
N.º de referencia del gen P30153, Q76MZ3
Alias de gen 29B; 6330556D22Rik; 65 kDa regulatory subunit of protein phosphatase; A/PR65; alpha isoform of regulatory subunit A, protein phosphatase 2; CG13383; CG17291; CG17291-PA; CG17291-PB; CG17291-PC; CG17291-PD; CG17291-PE; CG33297; Dmel\CG17291; Dmel_CG17291; dPP2A A; DPR65; Medium tumor antigen-associated 61 kDa protein; MGC786; MRD36; PP2A; PP2A 29B; PP2A 65kD subunit A; PP2A A; PP2A subunit A isoform PR65-alpha; PP2A subunit A isoform R1-alpha; PP2A[A]; Pp2A29B; PP2A-29B; Pp2A-29B-PA; Pp2A-29B-PB; Pp2A-29B-PC; Pp2A-29B-PD; Pp2A-29B-PE; PP2A-A; PP2AAALPHA; PP2A-Aalpha; PP2Ac; PP2CA; PP2Calpha; Ppp2r1a; PR65; PR65A; protein phosphatase 2 (formerly 2A), regulatory subunit A (PR 65), alpha isoform; protein phosphatase 2 (formerly 2A), regulatory subunit A , alpha isoform; protein phosphatase 2 (formerly 2A), regulatory subunit A, alpha isoform; protein phosphatase 2 regulatory subunit A, alpha; protein phosphatase 2 scaffold subunit Aalpha; protein phosphatase 2, regulatory subunit A, alpha; protein phosphatase 2A; protein phosphatase 2A at 29B; protein phosphatase PP2A; Protein phosphatase PP2A regulatory subunit A; protein phosphate 2A; regulatory subunit A of type 2A protein phosphatase; RP-C; serine/threonine protein phosphatase 2A, 65 kDa regulatory subunit A, alpha isoform; serine/threonine protein phosphatase A subunit type 2A; serine/threonine-protein phosphatase 2A 65 kDa regulatory subunit A alpha isoform; Serine/threonine-protein phosphatase PP2A 65 kDa regulatory subunit; testicular secretory protein Li 1; Unknown (protein for MGC:128399)
Símbolos de los genes PPP2R1A
Especie del huésped Rabbit
Inmunógeno Recombinant protein encompassing a sequence within the center region of human PPP2R1A. The exact sequence is proprietary.
Método de purificación Antigen affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 117281, 51792, 5518
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.