Learn More
Invitrogen™ PPME1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA527754
Descripción
Recommended positive controls: 293T, A431, HeLa, HepG2, NIH-3T3, JC, BCL-1. Predicted reactivity: Mouse (97%), Rat (97%), Zebrafish (81%), Xenopus laevis (86%), Chicken (91%), Rhesus Monkey (100%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Protein phosphatase methylesterase-1 catalyzes the demethylation of the protein phosphatase-2A catalytic subunit.
Especificaciones
| PPME1 | |
| Polyclonal | |
| Unconjugated | |
| Ppme1 | |
| 1110069N17Rik; 2700017M01Rik; Pme1; PME-1; PP2593; PPME1; PRO0750; Protein phosphatase methylesterase 1; protein phosphatase methylesterase-1; RGD1309683; testicular secretory protein Li 39 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51400, 72590 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q8BVQ5, Q9Y570 | |
| Ppme1 | |
| Recombinant protein encompassing a sequence within the center region of human PPME1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.