Descripción
This gene is a member of the cyclophilin family of peptidylprolyl isomerases. The cyclophilins are a highly conserved family, members of which play an important role in protein folding, immunosuppression by cyclosporin A, and infection of HIV-1 virions.
Specifications
Specifications
| Antígeno | PPIL4 |
| Aplicaciones | Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.26 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| génica | PPIL4 |
| N.º de referencia del gen | Q8WUA2, Q9CXG3 |
| Alias de gen | Cyclophilin like protein PPIL4, HDCME13P, PPIase, PPIL4, Rotamase PPIL4 |
| Símbolos de los genes | Ppil4 |
| Show More |