missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PPHLN1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA558583
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Immunogen sequence: KKPPLLDRPG EGSYNRYYSH VDYRDYDEGR SFSHDRRSGP PHRGDESGYR WTRDDHSASR QPEYRDMRDG FRRKSFYSSH YARERSPYKR DNTFFRESPV GRKDSPHSR Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 83%.
The protein encoded by this gene is one of the several proteins that become sequentially incorporated into the cornified cell envelope during the terminal differentiation of keratinocyte at the outer layers of epidermis. This protein interacts with periplakin, which is known as a precursor of the cornified cell envelope. The cellular localization pattern and insolubility of this protein suggest that it may play a role in epithelial differentiation and contribute to epidermal integrity and barrier formation. Multiple alternatively spliced transcript variants encoding distinct isoforms have been observed. [provided by RefSeq, Jul 2008].
Especificaciones
| PPHLN1 | |
| Polyclonal | |
| Unconjugated | |
| PPHLN1 | |
| CDC7 expression repressor; CR; Gastric cancer antigen Ga50; HSPC206; HSPC232; MGC48786; periphilin 1; periphilin-1; periphilin-1-like protein; Pphln1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51535 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8NEY8 | |
| PPHLN1 | |
| Recombinant protein corresponding to Human PPHLN1. Recombinant protein control fragment (Product #RP-97735). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido