Learn More
Invitrogen™ PPA1 Recombinant Rabbit Monoclonal Antibody (24GB1365)
GREENER_CHOICE
Código de producto. 30275393
Change view
Click to view available options
Cantidad:
20 μL
100 μL
Tamaño de la unidad:
100 microlitros
20 microlitros
30275393 100 μL 100 microlitros
30274993 20 μL 20 microlitros
2 options
Código de producto. 30275393
Marca: Invitrogen™ MA557430
Rabbit Recombinant Monoclonal Antibody
PPA1 Recombinant Monoclonal Antibody for Western Blot, Flow
The protein encoded by this gene is a member of the inorganic pyrophosphatase (PPase) family. PPases catalyze the hydrolysis of pyrophosphate to inorganic phosphate, which is important for the phosphate metabolism of cells. Studies of a similar protein in bovine suggested a cytoplasmic localization of this enzyme.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno PPA1
Aplicaciones Flow Cytometry, Western Blot, Immunocytochemistry
Clasificación Recombinant Monoclonal
Clon 24GB1365
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
génica Ppa1
N.º de referencia del gen Q15181, Q9D819
Alias de gen 2010317E03Rik; cytosolic inorganic pyrophosphatase; diphosphate phosphohydrolase; epididymis secretory sperm binding protein Li 66p; HEL-S-66p; inorganic diphosphatase; inorganic diphosphatase 1; inorganic pyrophosphatase; inorganic pyrophosphatase 1; IOPPP; Pp; PP1; Ppa1; PPase; Pyp; pyrophosphatase (inorganic) 1; pyrophosphatase 1; pyrophosphate phospho-hydrolase; RP11-367H5.1; SID6-8061
Símbolos de los genes Ppa1
Especie del huésped Rabbit
Inmunógeno A synthesized peptide derived from human Pyrophosphatase 1 (aa 1-45).
Método de purificación Affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 294504, 5464, 67895
Especies diana Human, Mouse, Rat
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG