Learn More
Brady™ Poste y bases rellenables
Código de producto. 11711306
Click to view available options
Color:
Blanco
Negro, amarillo
Rojo
Rojo, blanco
Tamaño de la unidad:
1 paquete
Código de producto. 11711306
Marca: Brady™ 256722
Solo quedan null
Añadir a la cesta
Añadir a la cesta
These Posts and Fillable Bases are ideal for controlling and directing foot traffic. Suitable for warehouses, factories, offices etc. Chains and ribbons can be attached to the sturdy 900 mm posts.
  • Ideal for controlling and directing foot traffic.
  • Suitable for warehouses, factories, offices etc.
  • Chains and ribbons can be attached to the sturdy 900 mm posts.
  • Posts and Fillable Bases.
Especificaciones
Alternate Part Number POST & FILLABLE BASE YELL/BLK PK 4
Diámetro exterior (métrico) 40 mm
Altura (imperial) 35.43 in.
Altura (métrico) 900 mm
Cantidad de embalaje 4 paquete
Color Negro, amarillo
Tipo Poste con base rellenable