Learn More
Sanplatec™ Portable Vacuum Sampling Box
Código de producto. 30245638
Change view
Click to view available options
Modelo:
CAJA DE 100 mL
CAJA DE 150 mL
CAJA DE 300 mL
CAJA DE 500 mL
CAJA DE 50 mL
RECUADRO Cuadrado
Tamaño de la unidad:
cada uno
Código de producto. Modelo unitSize
30245638 CAJA DE 300 mL cada uno
30245640 RECUADRO Cuadrado cada uno
30245631 CAJA DE 50 mL cada uno
30245648 CAJA DE 100 mL cada uno
30245633 CAJA DE 150 mL cada uno
30245641 CAJA DE 500 mL cada uno
6 options
Código de producto. 30245638
Marca: Sanplatec™ 19930
Solo quedan null
Añadir a la cesta
Añadir a la cesta
Mini size of sturdy acrylic desiccator with valves. Convenient to carry around.
  • This is an ultra-small size that is convenient to carry around.
  • Valves are equipped as standard, enabling gas purge by decompressing.
  • Check valves operate as relief valves. It is safe even if the inside of the box becomes positive pressure during gas purge.
  • Light weight (approx. 200 - 620 g).
  • It is convenient not only for laboratory sample storage but also for outdoor sampling (other example applications: sampling of soil, storage of trace samples).
  • Ball valve port: outer diameter 6 mm tube (one touch type).
  • Check valve connection port: outer diameter 6 mm tube (one touch type).
  • Check valve operating pressure: 10 kPa.
Especificaciones
Material Acrylic
Tipo de producto Vacuum Desiccator
Modelo CAJA DE 300 mL
Dimensiones interiores (F x An x Al) 48 x 186 mm
Altura (métrico) 246 mm
Diámetro interior (métrico) 48 mm
Diámetro exterior (imperial) 64 mm