Learn More
Kartell SpA Micropipette Tips
Descripción
Manufactured from virgin cadmium-free polypropylene. High clarity and excellent water-repellent properties designed to keep sample retention within the tip to a minimum. The precise centering of the orifice at the end of the tip guarantees reliable directionality of the sample when delivered.
Recommended for use with the Kartell automatic micropipettes but deliver excellent results even if used with the most other micropipette brands available on the market. Compatibility: Kartell™ , Gilson™, Eppendorf™ , Biohit™ Socorex™ , HTL™, Nichiryo™, Finnpipette™, Thermo™
Especificaciones
Especificaciones
| Volumen (métrico) | De 100 a 1000 μl |
| filtradas por | Non-filtered |
| Esterilidad | Non-sterile |
| Color | Azul |
| Esterilizables en autoclave | Sí |
| Certificaciones/conformidad | CE, IVD |
| Formato | Bagged |
| Estilo de punta | Punta de pipeta |
| Tipo de producto | Tips |
| Código de color | Light Blue |
| Mostrar más |