Learn More
Macherey-Nagel™ Placas de vidrio Polygram CEL 300-25
Product Code. 10114811 Shop All Macherey-Nagel Products
Change view
Click to view available options
Dimensiones (L x An):
20 x 20 cm
Unit Size:
paquete de 25
Product Code. Dimensiones (L x An) unitSize
10114811 20 x 20 cm paquete de 25
1 options
This item is not returnable. View return policy
Product Code. 10114811
Brand: Macherey-Nagel™ 808033
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
Cantidad 25/Paq.
Tipo Placa de vidrio
Grosor (métrico) 0,25 mm
Dimensiones (L x An) 20 x 20 cm