Learn More
Pal International™ Sobremangas de polietileno
Descripción
- High quality polyethylene construction
- Concealed elastic at the wrist and elbow
- Secure yet comfortable fit.
- Provides splash protection
- Ensure hygiene standards are met in a variety of industrial, manufacturing and healthcare environments.
- Suitable for use with food
- Latex-free to minimise the risk of irritation.
- Easily dispensed from the Pal Ecopak dispenser
Especificaciones
Especificaciones
| Tipo de producto | Cubremangas de uso general |
| Material | Polietileno |
| Color | Azul |
| Desechables | Single-use |
| Sin látex | Yes |