missing translation for 'onlineSavingsMsg'
Learn More

Polydatin, MedChemExpress

Código de producto. 16492346
Encompass_Preferred
Click to view available options
Cantidad:
100 mg
200 mg
500 mg
Tamaño de la unidad:
100 miligramos
200 miligramos
500 miligramos
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 16492346

Marca: Medchem Express HYN0120A

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

MedChemExpress Polydatin (Piceid), extracted from the roots of Polygonum cuspidatum Sieb, a widely used traditional Chinese remedies, possesses anti-inflammatory activity in several experimental models. Polydatin (Piceid) inhibits G6PD and induces oxidative and ER stresses.

Polydatin (Piceid), extracted from the roots of Polygonum cuspidatum Sieb, a widely used traditional Chinese remedies, possesses anti-inflammatory activity in several experimental models. Polydatin (Piceid) inhibits G6PD and induces oxidative and ER stresses.

Specifications

Nombre del producto químico o material Polydatin
CAS 27208-80-6
Color Blanco
Notas de grado de pureza Research
Fórmula molecular C20H22O8
Cantidad 500 mg
Sinónimo Piceid
Información de solubilidad DMSO : ≥ 29 mg/mL (74.29 mM) ∣H2O : 0.1 mg/mL (0.26 mM; Need ultrasonic)
SMILES O[C@H]([C@H]([C@@H]([C@@H](CO)O1)O)O)[C@@H]1OC2=CC(/C=C/C3=CC=C(O)C=C3)=CC(O)=C2.[(E)]
Peso molecular (g/mol) 390.38
Formula Weight (peso de la fórmula) 390.38
Porcentaje de pureza 98.55%
Grado Research
Forma física Solid
Para utilizar con (aplicación) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.