Learn More
Sanplatec™ Polycarbonate Vacuum Desiccator
Código de producto. p-200109728
Click to view available options
Diámetro interior (métrico):
200 mm
230 mm
Tamaño de la unidad:
cada uno
Código de producto. 30245647
Marca: Sanplatec™ 04705
Solo quedan null
Añadir a la cesta
Añadir a la cesta
Transparent polycarbonate with excellent visibility and airtightness as well as impact resistance and heat resistance. (with stopcock and gauge)
  • Clear view. Easy to see the condition of stored items and the timing of changing silica gel.
  • Dry seal type with O-ring. Worry-free of grease coating.
  • The highest intensity in transparent resin. Hardly broken when dropping.
  • The ceramic plate has heat resistance, corrosion resistance, and non-adhesiveness.
  • PC material feature: good pulling strength, shock strength, and heat-resistance
  • Spare plate for each type (PC-210/250 kg) is also available.
  • Nozzle diameter is for 9 mm.
Especificaciones
Material Polycarbonate
Tipo de producto Vacuum Desiccator
Incluye Stopcock, Gauge
Altura (métrico) 337 mm
Diámetro interior (métrico) 200 mm
Diámetro exterior (imperial) 220 mm