missing translation for 'onlineSavingsMsg'
Learn More

Poloxime, MedChemExpress

Código de producto. 16461979
Encompass_Preferred
Change view
Click to view available options
Cantidad:
5 mg
10 mg
50 mg
100 mg
500 mg
1 g
Tamaño de la unidad:
1 g
10 miligramos
100 miligramos
5 miligramos
50 miligramos
500 miligramos
Product Code. Cantidad unitSize
16461979 1 g 1 g
16464117 50 mg 50 miligramos
16457956 100 mg 100 miligramos
16451956 500 mg 500 miligramos
16410668 5 mg 5 miligramos
16408228 10 mg 10 miligramos
6 missing translation for 'options'
This item is not returnable. View return policy

Product Code. 16461979

missing translation for 'mfr': Medchem Express HY77195

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

MedChemExpress Poloxime, a hydrolysis product of poloxin, is a non-ATP-competitive Plk1 inhibitor, with moderate Plk1 inhibitory activity.

Poloxime, a hydrolysis product of poloxin, is a non-ATP-competitive Plk1 inhibitor, with moderate Plk1 inhibitory activity.

Specifications

Nombre del producto químico o material Poloxime
Forma física Solid
CAS 17302-61-3
Color Yellow
Cantidad 1 g
Notas de grado de pureza Research
Fórmula molecular C10H13NO2
Para utilizar con (aplicación) Cancer-Kinase/protease
Información de solubilidad DMSO : ≥ 100 mg/mL (557.97 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES CC1=CC(C(C(C)C)=C/C1=N\O)=O
Peso molecular (g/mol) 179.22
Formula Weight (peso de la fórmula) 179.22
Porcentaje de pureza 95.0%
Grado Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.