Learn More
MilliporeSigma™ Polishers for Milli-Q™ IQ 7000 Ultrapure Water System
Our Lab Water purification system portfolio offers a broad range of pure and ultrapure water purification systems and services designed for scientists working in pharmaceutical, clinical, academic, industrial, research, and government laboratories — in both validated and non-validated environments.
722.00€
Especificaciones
| Para utilizar con (equipo) | Milli-Q IQ 7000 Ultrapure Water System |
|---|---|
| Descripción | Delivered hermetically sealed |
| Código de producto | Marca | Tipo | Para utilizar con (aplicación) | Precio | Cantidad y disponibilidad | ||||
|---|---|---|---|---|---|---|---|---|---|
| Código de producto | Marca | Tipo | Para utilizar con (aplicación) | Precio | Cantidad y disponibilidad | ||||
|
15864792
|
MilliporeSigma™
CDUFBI0A1 |
Biopak™ Polisher | Ultrafilter for the production of pyrogen-, nuclease-, protease- and bacteria-free water at the point of dispense of Milli-Q IQ 7000 system |
722.00€
cada uno |
Fecha estimada de envoi: 29-01-2026
Inicie sesión para ver el stock |
Inicie sesión o regístrese para comprobar su precio y disponibilidad.
|
|||
Descripción
Our Lab Water purification system portfolio offers a broad range of pure and ultrapure water purification systems and services designed for scientists working in pharmaceutical, clinical, academic, industrial, research, and government laboratories — in both validated and non-validated environments.
For use with Milli-Q™ IQ 7000 Ultrapure Water System.Especificaciones
| Milli-Q IQ 7000 Ultrapure Water System | |
| Delivered hermetically sealed |