missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
This is a paternally expressed imprinted gene that encodes transcripts containing two overlapping open reading frames start codon, have been reported for this gene. Increased expression of this gene is associated with hepatocellular carcinomas.
Especificaciones
Especificaciones
| Antígeno | POLI |
| Aplicaciones | Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.18 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| génica | POLI |
| N.º de referencia del gen | Q6R3M4, Q9UNA4 |
| Alias de gen | DNA polymerase iota, Eta2, POLI, polymerase (DNA directed) iota, RAD30 homolog B, RAD30B, RAD3OB |
| Símbolos de los genes | Poli |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?