Learn More
Podocalyxin Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca:  Proteintech 18150-1-AP-150UL
 Ver más versiones de este producto
Código de producto. 16828204
  • 410.00€ / 150 microlitros
Descripción
PODXL encodes a member of the sialomucin protein family. The encoded protein was originally identified as an important component of glomerular podocytes. Podocytes are highly differentiated epithelial cells with interdigitating foot processes covering the outer aspect of the glomerular basement membrane. Other biological activities of the encoded protein include: binding in a membrane protein complex with Na+/H+ exchanger regulatory factor to intracellular cytoskeletal elements, playing a role in hematopoetic cell differentiation, and being expressed in vascular endothelium cells and binding to L-selectin.
Especificaciones
Especificaciones
Podocalyxin
Polyclonal
Unconjugated
PODXL
GCTM 2 antigen, Gp200, PC, PCLP, PCLP 1, PCLP1, Podocalyxin, podocalyxin like, Podocalyxin like protein 1, PODXL, RCC marker, Renal Cell Carcinoma marker, TRA 1 81,TRA 1 60, TRA-1-60
Rabbit
Antigen Affinity Chromatography
RUO
27205, 5420
-20°C
Liquid
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
0.26 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O00592, Q9R0M4
Podxl
Podocalyxin Fusion Protein Ag12844
150 μL
Primary
Human, Mouse